Learn More
National Diagnostics ProtoGlow ECL | 200mL
SDP
Catalog No. NC1478047
Encompass
ProtoGlow ECL delivers the latest technology developed for enhanced chemiluminescent detection on Western blots. The unique chemistry of the ProtoGlow ECL system increases the sensitivity of Western blots by up to 20 fold. This allows the detection of proteins at lower abundance and/or the use of higher dilutions of primary and secondary antibodies, economizing on these expensive reagents.
Catalog No. NC1478047 Supplier National Diagnostics Supplier No. CL300200ML
ProtoGlow ECL delivers the latest technology developed for enhanced chemiluminescent detection on Western blots. The unique chemistry of the ProtoGlow ECL system increases the sensitivity of Western blots by up to 20 fold. This allows the detection of proteins at lower abundance and/or the use of higher dilutions of primary and secondary antibodies, economizing on these expensive reagents.