Learn More
Medchemexpress LLC Protogracillin | 54848-30-5 | 1 MG
SDP
Catalog No. 5000420945
Encompass_Preferred
Protogracillin is a steroidal saponin isolated from the plant Dioscorea zingiberensis Wright. Steroidal saponins found in the rhizomes of this plant have the potential to reduce the risk of cardiovascular diseases through anti-thrombotic action. It is intended for research use only and not for sale to patients.
  • It is a steroidal saponin.
  • It is isolated from Dioscorea zingiberensis Wright.
  • It has potential anti-thrombotic action.
  • It has shown cytotoxicity against human CCRF-CEM cells with an IC50 of 1.77 μM.
  • Its purity is 99.00%.
Catalog No. 50-004-20945 Supplier Medchemexpress LLC Supplier No. HYN42711MG
Protogracillin is a steroidal saponin isolated from the plant Dioscorea zingiberensis Wright. Steroidal saponins found in the rhizomes of this plant have the potential to reduce the risk of cardiovascular diseases through anti-thrombotic action. It is intended for research use only and not for sale to patients.
  • It is a steroidal saponin.
  • It is isolated from Dioscorea zingiberensis Wright.
  • It has potential anti-thrombotic action.
  • It has shown cytotoxicity against human CCRF-CEM cells with an IC50 of 1.77 μM.
  • Its purity is 99.00%.