Learn More
TARGETMOL CHEMICALS INC PROTOHYPERICIN 5MG
SDP
Catalog No. 502578882
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Protohypericin is a naturally occurring naphthodianthrone derived from plant Hypericum perforatum.Radioiodinated protohypericin is used in Tumor necrosis targeted radiotherapy. Protohypericin exhibits photocytotoxicity. purity: 98%
Catalog No. 50-257-8882 Supplier TARGETMOL CHEMICALS INC Supplier No. TN21085MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Protohypericin is a naturally occurring naphthodianthrone derived from plant Hypericum perforatum.Radioiodinated protohypericin is used in Tumor necrosis targeted radiotherapy. Protohypericin exhibits photocytotoxicity. purity: 98%