Learn More
ZORO TOOLS PROTOJ000AA25BLDEFEELERGAUGEST
Catalog No. NC1223876
Encompass
Short Blade Feeler Gauge Set, Range (In.) 0.0015 to 0.040, Size of Leaves 1/2 x 3 In., Number of Leaves 25, 0.0015, 0.002, 0.003, 0.004, 0.005, 0.006, 0.007, 0.008, 0.009, 0.010, 0.011, 0.012, 0.013, 0.014, 0.015, 0.016, 0.017, 0.018, 0.020, 0.022, 0.025, 0.030, 0.032, 0.035, 0.040, Includes Steel Holder
Catalog No. NC1223876 Supplier ZORO TOOLS Supplier No. G2259275
Short Blade Feeler Gauge Set, Range (In.) 0.0015 to 0.040, Size of Leaves 1/2 x 3 In., Number of Leaves 25, 0.0015, 0.002, 0.003, 0.004, 0.005, 0.006, 0.007, 0.008, 0.009, 0.010, 0.011, 0.012, 0.013, 0.014, 0.015, 0.016, 0.017, 0.018, 0.020, 0.022, 0.025, 0.030, 0.032, 0.035, 0.040, Includes Steel Holder