Learn More
Apexbio Technology LLC Protoporphyrin IX, 100mg CAS# 553-12-8
SDP
Catalog No. 501998252
Encompass_Preferred
Protoporphyrin IX is a member of porphyrin family, which belongs to tetrapyrroles. It is the precursor of heme in biosynthetic pathway. Protoporphyrin IX can be used in protoporphyrin assays, fluorescence spectra analysis and heme-mediated ferroportin 1 transcription study. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8252 Supplier Apexbio Technology LLC Supplier No. B8225100
Protoporphyrin IX is a member of porphyrin family, which belongs to tetrapyrroles. It is the precursor of heme in biosynthetic pathway. Protoporphyrin IX can be used in protoporphyrin assays, fluorescence spectra analysis and heme-mediated ferroportin 1 transcription study. Other sizes are also available. Please inqury us for quote.