Learn More
Selleck Chemical LLC Protoporphyrin IX (PPIX) S5145-25mg
SDP
Catalog No. 503111980
Encompass_Preferred
Protoporphyrin IX (PPIX) is a heterocyclic organic compound which consists of four pyrrole rings and is the final intermediate in the heme biosynthetic pathway
Catalog No. 50-311-1980 Supplier Selleck Chemical LLC Supplier No. S514525MG
Protoporphyrin IX (PPIX) is a heterocyclic organic compound which consists of four pyrrole rings and is the final intermediate in the heme biosynthetic pathway