Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protoporphyrin IX | 553-12-8 | 96.1% | 562.66 g/mol | C34H34N4O4 | 10 MG
SDP

Catalog No. 5000297456
Encompass_Preferred

Protoporphyrin IX analytical standard for research and analytical applications, commonly used in studies of heme biosynthesis, photodynamic and sonodynamic agents, and fluorescence-based assays. Supplied as a small laboratory pack suitable for analytical testing.

  • Analytical standard with reported purity of 96.09%.
  • Molecular weight 562.66 g/mol and formula C34H34N4O4.
  • Provided in a 10 mg laboratory pack for assay and method validation.
  • Suitable for photodynamic and sonodynamic research and fluorescence assays.
  • Certificate of analysis and safety datasheet available for quality and handling information.

Catalog No. 50-002-97456 Supplier Medchemexpress LLC Supplier No. HYB1247R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protoporphyrin IX analytical standard for research and analytical applications, commonly used in studies of heme biosynthesis, photodynamic and sonodynamic agents, and fluorescence-based assays. Supplied as a small laboratory pack suitable for analytical testing.

  • Analytical standard with reported purity of 96.09%.
  • Molecular weight 562.66 g/mol and formula C34H34N4O4.
  • Provided in a 10 mg laboratory pack for assay and method validation.
  • Suitable for photodynamic and sonodynamic research and fluorescence assays.
  • Certificate of analysis and safety datasheet available for quality and handling information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.