Learn More
Medchemexpress LLC Protoporphyrin IX | 553-12-8 | MFCD00151109 | 98.0% | 100 MG
SDP
Catalog No. 5000412897
Encompass_Preferred
Protoporphyrin IX is an endogenous metabolite and a final intermediate in the heme biosynthetic pathway. It functions as a radiation sensitizer, enhancing ROS generation even in hypoxic conditions and inducing DNA damage. It also acts as a photosensitizer that undergoes photobleaching from light irradiation. It accumulates in tumor cells after administration of 5-aminolevulinic acid (5-ALA).
  • Endogenous metabolite and final intermediate in heme biosynthetic pathway
  • Functions as a radiation sensitizer
  • Enhances ROS generation even in hypoxic conditions
  • Induces DNA damage
  • Acts as a photosensitizer that undergoes photobleaching from light irradiation
  • Accumulates in tumor cells after administration of 5-aminolevulinic acid (5-ALA)
  • Shows promise as a sonodynamic and photodynamic agent for various cancers, such as bladder cancer and nodular basal cell carcinoma
Catalog No. 50-004-12897 Supplier Medchemexpress LLC Supplier No. HYB1247R100MG
Protoporphyrin IX is an endogenous metabolite and a final intermediate in the heme biosynthetic pathway. It functions as a radiation sensitizer, enhancing ROS generation even in hypoxic conditions and inducing DNA damage. It also acts as a photosensitizer that undergoes photobleaching from light irradiation. It accumulates in tumor cells after administration of 5-aminolevulinic acid (5-ALA).
  • Endogenous metabolite and final intermediate in heme biosynthetic pathway
  • Functions as a radiation sensitizer
  • Enhances ROS generation even in hypoxic conditions
  • Induces DNA damage
  • Acts as a photosensitizer that undergoes photobleaching from light irradiation
  • Accumulates in tumor cells after administration of 5-aminolevulinic acid (5-ALA)
  • Shows promise as a sonodynamic and photodynamic agent for various cancers, such as bladder cancer and nodular basal cell carcinoma