Learn More
Medchemexpress LLC Protoporphyrin IX (Standard) | 553-12-8 | 96.1% | 100 MG
SDP
Catalog No. 5000413224
Encompass_Preferred
Protoporphyrin IX (Standard) is the analytical standard for Protoporphyrin IX. It is intended for research and analytical applications, serving as a final intermediate in the heme biosynthetic pathway. This compound acts as a radiation sensitizer, enhancing ROS generation, and induces DNA damage. It also functions as a photosensitizer, undergoing photobleaching through direct light degradation. Protoporphyrin IX is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for Protoporphyrin IX
  • Acts as a radiation sensitizer and photosensitizer
  • Used in HPLC, GC, and MS research experiments
  • Suitable for research of sonodynamic and photodynamic agents for various cancers
Catalog No. 50-004-13224 Supplier Medchemexpress LLC Supplier No. HYB1247A100MG
Protoporphyrin IX (Standard) is the analytical standard for Protoporphyrin IX. It is intended for research and analytical applications, serving as a final intermediate in the heme biosynthetic pathway. This compound acts as a radiation sensitizer, enhancing ROS generation, and induces DNA damage. It also functions as a photosensitizer, undergoing photobleaching through direct light degradation. Protoporphyrin IX is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for Protoporphyrin IX
  • Acts as a radiation sensitizer and photosensitizer
  • Used in HPLC, GC, and MS research experiments
  • Suitable for research of sonodynamic and photodynamic agents for various cancers