Learn More
Selleck Chemical LLC Protoporphyrin IX Zinc
SDP
Catalog No. 5000003599
Encompass_Preferred
Zinc Protoporphyrin (Zn(II)-protoporphyrin IX) is an potent orally active and competitive heme oxygenase-1 (HO-1) inhibitor and significantly reduces the protective effects of Phloroglucinol (PG) against hydrogen peroxide This product has poor solubility animal experiments are available cell experiments please choose carefully
Catalog No. 50-000-03599 Supplier Selleck Chemical LLC Supplier No. S788025MG
Zinc Protoporphyrin (Zn(II)-protoporphyrin IX) is an potent orally active and competitive heme oxygenase-1 (HO-1) inhibitor and significantly reduces the protective effects of Phloroglucinol (PG) against hydrogen peroxide This product has poor solubility animal experiments are available cell experiments please choose carefully