Learn More
Selleck Chemical LLC Protoporphyrin IX Zinc-5mg
SDP
Supplier:  Selleck Chemical LLC S78805MG
Encompass_Preferred
Zinc Protoporphyrin (Zn(II)-protoporphyrin IX) is an potent, orally active and competitive heme oxygenase-1 (HO-1) inhibitor and significantly reduces the protective effects of Phloroglucinol (PG) against hydrogen peroxide.This product has poor solubility, animal experiments are available, cell experiments please choose carefully!
Catalog No. 50-238-9871