Learn More
Enzo Life Sciences Protoporphyrin IX (free acid) (1g). CAS: 553-12-8
SDP
Catalog No. 502009166
Encompass_Preferred
Protoporphyrin IX free acid, as distinct from its zinc salt (which inhibits heme oxygenase), activates soluble guanylyl cyclase (sGC) by binding directly to the enzyme. A useful reagent in cases where the use of nitric oxide (NO) or nitric oxide donors is undesirable. Alternative name: 8,13-Divinyl-3,7,12,17-tetramethyl-21H,23H-porphine-2,18-dipropionic acid. Purity: ≥95% (HPLC). Appearance: Purple to dark brown solid. Solubility: Dissolve initially in 0.1M base (Tris base or NaOH), then add water miscible organic solvent (EtOH (10mg/ml), MeOH, DMSO (10mg/ml), DMF) until solution is 50/50 base/solvent. When a clear solution is obtained, it can be diluted into an aqueous medium and titrated or buffered to any pH>7. Use immediately. Long Term Storage: -20°C.
Catalog No. 50-200-9166 Supplier Enzo Life Sciences Supplier No. ALX430041G001
Protoporphyrin IX free acid, as distinct from its zinc salt (which inhibits heme oxygenase), activates soluble guanylyl cyclase (sGC) by binding directly to the enzyme. A useful reagent in cases where the use of nitric oxide (NO) or nitric oxide donors is undesirable. Alternative name: 8,13-Divinyl-3,7,12,17-tetramethyl-21H,23H-porphine-2,18-dipropionic acid. Purity: ≥95% (HPLC). Appearance: Purple to dark brown solid. Solubility: Dissolve initially in 0.1M base (Tris base or NaOH), then add water miscible organic solvent (EtOH (10mg/ml), MeOH, DMSO (10mg/ml), DMF) until solution is 50/50 base/solvent. When a clear solution is obtained, it can be diluted into an aqueous medium and titrated or buffered to any pH>7. Use immediately. Long Term Storage: -20°C.