Learn More
Synbiosis™ Protos 3 Instrument Validation Procedures
GREENER_CHOICESDP
Catalog No. 01257328 Shop All Synoptics, Inc. Products
Catalog No. 01-257-328 Supplier Synbiosis™ Supplier No. IQOQPQPS3
Add to Cart
Add to Cart
Protos 3 Instrument Validation Procedures take the user through each step in the verification of a Synbiosis™ system regulatory compliances and standard operational procedures (SOP). These simple procedures allow the user to verify that a system meets defined installation, operational, and performance quality specifications.
  • Installation Qualification (IQ): Verifies that when a system is unpacked it conforms with all purchasing, shipping, and unpacking documents. Once this procedure has been followed the system is then ready for the operational check process.
  • Operational Qualification (OQ): Establishes if the system meets all the operation and functionality specifications. An operational test is also included at this stage.
  • Performance Qualification (PQ): Establishes if the system meets the specification for the nature of the purpose it was intended for. This is verified by using a simple target to establish if the system can meet performance criteria.
  • Documentation: Each set of IQ/OQ/PQ documents contains a complete set of procedures and checklists for following the verification steps. Included with each pack is a simple calibration device (target image) so that the performance of the system can be checked against the PQ requirements. Each set of documents is unique to a particular system and has to be ordered with the system itself. The document pack has full instructions for use and implementation.
Specifications
For Use With (Application) Instrument Validation
WARNING: Cancer - www.P65Warnings.ca.gov