Learn More
Medchemexpress LLC HY-N0800 5mg Medchemexpress, Protosappanin B CAS:102036-29-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN08005MG
Encompass_Preferred
Medchemexpress, HY-N0800 5mg Protosappanin B CAS:102036-29-3 Protosappanin B is a phenolic compound extracted from Lignum Sappan. Anti-cancer activity. Protosappanin B induces apoptosis and causes G1 cell cycle arrest in human bladder cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0310