Learn More
Andwin Scientific PROTOSLO QUIETING SOLUTION
SDP
Catalog No. NC9548699
Encompass_Preferred
The Protoslo Quieting Solution is made to decelerate the movement of protozoa to keep them in focus and in the field of view while also keeping their characteristic motion. This clear, colorless solution is packaged and sold in a 15 mL dropper bottle.
Catalog No. NC9548699 Supplier Andwin Scientific Supplier No. 885141
The Protoslo Quieting Solution is made to decelerate the movement of protozoa to keep them in focus and in the field of view while also keeping their characteristic motion. This clear, colorless solution is packaged and sold in a 15 mL dropper bottle.