Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protostemotinine | 169534-85-4 | 99.4% | 415.48 | C23H29NO6 | 10 MG
SDP

Catalog No. 5000295902
Encompass_Preferred

Protostemotinine is a natural alkaloid isolated from the roots and rhizomes of Stemona sessilifolia and supplied for research use. Analytical data and safety information are provided to support laboratory handling and small-scale experiments.

  • Natural alkaloid isolated from Stemona sessilifolia.
  • High purity (99.4%) with verified analytical data.
  • Molecular formula C23H29NO6; molecular weight 415.48.
  • CAS number 169534-85-4 for unambiguous identification.
  • Soluble in DMSO (25 mg/mL); sonication recommended.
  • COA and SDS available for download.
  • Packaged as 10 mg for small-scale research use.

Catalog No. 50-002-95902 Supplier Medchemexpress LLC Supplier No. HYN195510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protostemotinine is a natural alkaloid isolated from the roots and rhizomes of Stemona sessilifolia and supplied for research use. Analytical data and safety information are provided to support laboratory handling and small-scale experiments.

  • Natural alkaloid isolated from Stemona sessilifolia.
  • High purity (99.4%) with verified analytical data.
  • Molecular formula C23H29NO6; molecular weight 415.48.
  • CAS number 169534-85-4 for unambiguous identification.
  • Soluble in DMSO (25 mg/mL); sonication recommended.
  • COA and SDS available for download.
  • Packaged as 10 mg for small-scale research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.