Learn More
3B Scientific™ Protozoa Slide Set
SDPSCI_ED
Catalog No. S95032L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
10
Catalog No. Quantity
S95032L 10
1 options
Catalog No. S95032L Supplier 3B Scientific™ Supplier No. W13030
Add to Cart
Add to Cart
10 microscope slides
  • 1(e). Amoeba proteus, Rhizopoda, w.m.
  • 2(d). Radiolaria, mixed species, fossil
  • 3(d). Foraminifera from Mediterranean sea, mixed species, recent
  • 4(c). Euglena viridis, a green flagellate, w.m.
  • 5(c). Ceratium hirundinella, fresh-water dinoflagellate w.m.
  • 6(f). Trypanosoma gambiense, causes African sleeping sickness, blood smear
  • 7(f). Plasmodium, causes human malaria, blood smear
  • 8(d). Eimeria stiedae, causing coccidiosis, t.s. of infected liver
  • 9(d). Paramecium, a common ciliate, nuclei stained
  • 10(e). Vorticella, a coloniate ciliate
Specifications
Type Protozoa
Quantity 10