Learn More
TARGETMOL CHEMICALS INC Protriptyline hydrochloride
SDP
Catalog No. 502880514
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Protriptyline Hydrochloride is the hydrochloride salt form of protriptyline, a tricyclic secondary amine with an antidepressant property. Protriptyline hydrochloride (Protriptyline HCl) blocks the re-uptake of norepinephrine and serotonin by nerve terminals, thereby increasing available norepinephrine and serotonin. In addition, this agent exhibits anticholinergic activity. Purity 99.98%
Catalog No. 50-288-0514 Supplier TARGETMOL CHEMICALS INC Supplier No. T020450MG
Also available in 1 mL, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Protriptyline Hydrochloride is the hydrochloride salt form of protriptyline, a tricyclic secondary amine with an antidepressant property. Protriptyline hydrochloride (Protriptyline HCl) blocks the re-uptake of norepinephrine and serotonin by nerve terminals, thereby increasing available norepinephrine and serotonin. In addition, this agent exhibits anticholinergic activity. Purity 99.98%