Learn More
Medchemexpress LLC Protriptyline hydrochloride | 1225-55-4 | 1 ML
SDP
Catalog No. 5000408133
Encompass_Preferred
Protriptyline hydrochloride is a potent tricyclic antidepressant (TCA) that inhibits Acetylcholinesterase (AChE) activity and Aβ self-assembly. It is used in the study of depression and Alzheimer's disease. In vitro, it causes cytotoxicity in PC3 cells. In vivo, it has been shown to improve spatial learning and retention memory in rat models of Alzheimer's disease.
  • Inhibits AChE activity with an IC50 value of 0.06 mM.
  • Inhibits Aβ self-assembly.
  • Causes cytotoxicity in PC3 cells.
  • Improves spatial learning and retention memory in rat models of Alzheimer's disease.
Catalog No. 50-004-08133 Supplier Medchemexpress LLC Supplier No. HYB094910MM/1ML
Protriptyline hydrochloride is a potent tricyclic antidepressant (TCA) that inhibits Acetylcholinesterase (AChE) activity and Aβ self-assembly. It is used in the study of depression and Alzheimer's disease. In vitro, it causes cytotoxicity in PC3 cells. In vivo, it has been shown to improve spatial learning and retention memory in rat models of Alzheimer's disease.
  • Inhibits AChE activity with an IC50 value of 0.06 mM.
  • Inhibits Aβ self-assembly.
  • Causes cytotoxicity in PC3 cells.
  • Improves spatial learning and retention memory in rat models of Alzheimer's disease.