Learn More
Medchemexpress LLC Protriptyline hydrochloride | 1225-55-4 | 299.84 | 50 MG
SDP
Supplier:  Medchemexpress LLC 50MGHYB0949
Encompass_Preferred
Protriptyline hydrochloride is a potent tricyclic antidepressant (TCA). It inhibits AChE activity with an IC50 value of 0.06 mM and inhibits Aβ self-assembly. It can be used for the study of depression and Alzheimer's disease.
  • Potent tricyclic antidepressant (TCA)
  • Inhibits AChE activity with an IC50 value of 0.06 mM
  • Inhibits Aβ self-assembly
  • Suitable for studies on depression and Alzheimer's disease
  • Demonstrates cytotoxicity in PC3 cells (in vitro)
  • Improves spatial learning and retention memory in STZ treated rats (in vivo)
Catalog No. 50-003-64258