Learn More
Medchemexpress LLC Protriptyline hydrochloride | 1225-55-4 | 99.75% | 5 MG
SDP
Catalog No. 5000408833
Encompass_Preferred
Protriptyline hydrochloride is a potent tricyclic antidepressant (TCA) that inhibits AChE activity with an IC50 value of 0.06 mM and inhibits Aβ self-assembly. It can be used for the study of depression and Alzheimer's disease.
  • Inhibits AChE activity with an IC50 value of 0.06 mM
  • Inhibits Aβ self-assembly
  • Causes cytotoxicity in PC3 cells
  • Improves spatial learning and retention memory in STZ treated rats
  • Reduces pTau, Aβ42 and BACE-1 levels, neurodegeneration, oxidative stress, and glial activation
  • Improves p-ERK/ERK ratio and enhances BDNF and CREB levels by reducing NFκB and GFAP expression
Catalog No. 50-004-08833 Supplier Medchemexpress LLC Supplier No. HYB09495MG
Protriptyline hydrochloride is a potent tricyclic antidepressant (TCA) that inhibits AChE activity with an IC50 value of 0.06 mM and inhibits Aβ self-assembly. It can be used for the study of depression and Alzheimer's disease.
  • Inhibits AChE activity with an IC50 value of 0.06 mM
  • Inhibits Aβ self-assembly
  • Causes cytotoxicity in PC3 cells
  • Improves spatial learning and retention memory in STZ treated rats
  • Reduces pTau, Aβ42 and BACE-1 levels, neurodegeneration, oxidative stress, and glial activation
  • Improves p-ERK/ERK ratio and enhances BDNF and CREB levels by reducing NFκB and GFAP expression