Learn More
Medchemexpress LLC Protriptyline hydrochloride | 1225-55-4 | 99.8% | 25 MG
SDP
Catalog No. 5000450970
Encompass_Preferred
Protriptyline hydrochloride is a potent tricyclic antidepressant (TCA) that inhibits AChE activity with an IC50 value of 0.06 mM and also inhibits Aβ self-assembly. It is used for the study of depression and Alzheimer's disease.
  • Potent tricyclic antidepressant (TCA)
  • Inhibits AChE activity
  • Inhibits Aβ self-assembly
  • Causes cytotoxicity in PC3 cells
  • Improves spatial learning and retention memory in AD rat models
  • Reduces pTau, Aβ42, and BACE-1 levels
  • Enhances BDNF and CREB levels
Catalog No. 50-004-50970 Supplier Medchemexpress LLC Supplier No. 25MGHYB0949
Protriptyline hydrochloride is a potent tricyclic antidepressant (TCA) that inhibits AChE activity with an IC50 value of 0.06 mM and also inhibits Aβ self-assembly. It is used for the study of depression and Alzheimer's disease.
  • Potent tricyclic antidepressant (TCA)
  • Inhibits AChE activity
  • Inhibits Aβ self-assembly
  • Causes cytotoxicity in PC3 cells
  • Improves spatial learning and retention memory in AD rat models
  • Reduces pTau, Aβ42, and BACE-1 levels
  • Enhances BDNF and CREB levels