Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Pre-programmed to work with more than 150 Spectroquant™ test kits, Spectroquant™ Prove plus photometers are used for analysis.
- Enhanced functionality for operation and responsiveness
- Improved display responsiveness and faster processing
- Storage capacity for up to 7000 standard measurement results
- Warranty period of 2 years
- Easy to use and pre-programmed with reagents
- Reduced use of chemicals compared to traditional methods
- Suitable for a broad measuring range per parameter (minimizes dilution work)
- Pre-programmed for over 300 analysis methods
- Instrument-supported AQA system for quality assurance
- Automatic cell size recognition for cell and reagent tests
- Live ID barcode for all cell and reagent tests (including lot and expiry data)
- No adapter needed for measurement of different cell sizes
- Kits can be stored at room temperature
Specifications
Specifications
| Detector Type | Silicon Photodiode |
| Display | Touchscreen |
| Languages | English, German, French, Spanish, Italian |
| Light Source | Xenon Flash |
| Spectral Bandwidth | 4 nm |
| Voltage | 24 VDC |
| Wavelength Range | 190 to 1100 nm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?