Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supelco Prove 600 Plus

Catalog No. M1730280001 Shop All MilliporeSigma Products
Change view
Click to view available options
Wavelength Range:
190 to 1100 nm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength Range
M1730280001 190 to 1100 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1730280001 Supplier Supelco Supplier No. 1.73028.0001
Add to Cart
Add to Cart

Photometric analysis using Spectroquant™, covering a wavelength range of 190 to 1100 nm.

Pre-programmed to work with more than 150 Spectroquant™ test kits, Spectroquant™ Prove plus photometers are used for analysis.

  • Enhanced functionality for operation and responsiveness
  • Improved display responsiveness and faster processing
  • Storage capacity for up to 7000 standard measurement results
  • Warranty period of 2 years
  • Easy to use and pre-programmed with reagents
  • Reduced use of chemicals compared to traditional methods
  • Suitable for a broad measuring range per parameter (minimizes dilution work)
  • Pre-programmed for over 300 analysis methods
  • Cell size up to 10 cm with Prove 600 plus for high-sensitivity measurements
  • Instrument-supported AQA system for quality assurance
  • Automatic cell size recognition for cell and reagent tests
  • Live ID barcode for all cell and reagent tests (including lot and expiry data)
  • No adapter needed for measurement of different cell sizes
  • Kits can be stored at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Detector Type Silicon Photodiode
Display Touchscreen
Languages English, German, French, Spanish, Italian
Light Source Xenon Flash
Spectral Bandwidth 1.8 nm
Voltage 24 VDC
Wavelength Range 190 to 1100 nm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.