Learn More
ILEX LIFE SCIENCES LLC Provis Biolabs PNGase F Recombinant E coli expressed Elizabethkingia miricola enzyme 500 U 500 UN 1/EA
SDP
Catalog No. 5000580276
Encompass_Preferred
Provis Biolabs PNGase F Recombinant (cloned from Elizabethkingia miricola) RUO Grade 500 U DESCRIPTION PNGase F is an endoglycosidase that removes N-linked glycan oligosaccharides from glycoproteins Manufactured under ISO 9001 2015 Animal-origin-free (AOF) Research use only (RUO) GMP Grade is available upon request (made-to-order custom production) APPLICATION Glycoprotein analysis Protein structure determination Antibody deglycosylation Amino acids sequence determination X-Ray crystallography MALDI-TOF or other mass-spec techniques CAS 83534-39-8 SOURCE E coli recombinant (6xHis-tagged) MASS 36 kDa PURITY (SDS-PAGE) 95% FORMULATION 10 000 U/mL in 20 mM Tris 50 mM NaCl 5 mM EDTA (0 2 m filtered) ACTIVITY 1 U deglycosylation of 1 nmol RNase B/min at 37 C STORAGE 2-8 C MANUFACTURER Provis Biolabs ORIGIN India OTHER NAMES PNGase F enzyme PNGaseF Glycopeptide N-glycosidase N-glycanase N-Glycosidase F Peptide N-glycosidase EC 3 5 1 52
Catalog No. 50-005-80276 Supplier ILEX LIFE SCIENCES LLC Supplier No. PB06500U
Provis Biolabs PNGase F Recombinant (cloned from Elizabethkingia miricola) RUO Grade 500 U DESCRIPTION PNGase F is an endoglycosidase that removes N-linked glycan oligosaccharides from glycoproteins Manufactured under ISO 9001 2015 Animal-origin-free (AOF) Research use only (RUO) GMP Grade is available upon request (made-to-order custom production) APPLICATION Glycoprotein analysis Protein structure determination Antibody deglycosylation Amino acids sequence determination X-Ray crystallography MALDI-TOF or other mass-spec techniques CAS 83534-39-8 SOURCE E coli recombinant (6xHis-tagged) MASS 36 kDa PURITY (SDS-PAGE) 95% FORMULATION 10 000 U/mL in 20 mM Tris 50 mM NaCl 5 mM EDTA (0 2 m filtered) ACTIVITY 1 U deglycosylation of 1 nmol RNase B/min at 37 C STORAGE 2-8 C MANUFACTURER Provis Biolabs ORIGIN India OTHER NAMES PNGase F enzyme PNGaseF Glycopeptide N-glycosidase N-glycanase N-Glycosidase F Peptide N-glycosidase EC 3 5 1 52