Learn More
ILEX LIFE SCIENCES LLC Provis Biolabs rHA Recombinant Human Albumin Expressed in Pichia pastoris yeast 10 g 10 GR 1/EA
SDP
Catalog No. 5000580203
Encompass_Preferred
Provis Biolabs Recombinant Human Albumin (rHA) Expressed in Pichia pastoris Lyophilized Powder RUO Grade 10 g DESCRIPTION Provis Biolabs Recombinant Human Albumin (rHA) expressed in Pichia pastoris is a synthetic alternative to human serum albumin (HSA) The non-glycosylated polypeptide chain contains 585 amino acids and has a molecular mass of 67 kDa It does not contain any animal-derived components thus offering a reliable and sustainable alternative to traditional HSA sources Manufactured under ISO 9001 2015 Animal-origin-free (AOF) Research use only (RUO) GMP Grade is available upon request CAS 70024-90-7 SOURCE Protein is expressed in Pichia pastoris yeast (animal-free non-glycosylated) APPEARANCE An off-white to pale yellow powder PURITY 98% (SDS-PAGE HPLC) ENDOTOXIN 0 125 EU/mg STORAGE 2-8 C OPTIMAL CONCENTRATION (CELL CULTURE) 0 5 2 0 g/L MANUFACTURER Provis Biolabs pvt Ltd COUNTRY OF ORIGIN India
Catalog No. 50-005-80203 Supplier ILEX LIFE SCIENCES LLC Supplier No. PB03010G
Provis Biolabs Recombinant Human Albumin (rHA) Expressed in Pichia pastoris Lyophilized Powder RUO Grade 10 g DESCRIPTION Provis Biolabs Recombinant Human Albumin (rHA) expressed in Pichia pastoris is a synthetic alternative to human serum albumin (HSA) The non-glycosylated polypeptide chain contains 585 amino acids and has a molecular mass of 67 kDa It does not contain any animal-derived components thus offering a reliable and sustainable alternative to traditional HSA sources Manufactured under ISO 9001 2015 Animal-origin-free (AOF) Research use only (RUO) GMP Grade is available upon request CAS 70024-90-7 SOURCE Protein is expressed in Pichia pastoris yeast (animal-free non-glycosylated) APPEARANCE An off-white to pale yellow powder PURITY 98% (SDS-PAGE HPLC) ENDOTOXIN 0 125 EU/mg STORAGE 2-8 C OPTIMAL CONCENTRATION (CELL CULTURE) 0 5 2 0 g/L MANUFACTURER Provis Biolabs pvt Ltd COUNTRY OF ORIGIN India