Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROVON™ Citrus Ginger Foam Hand and Shower Wash
GREENER_CHOICESDP

Catalog No. 19043958 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
1250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-043-958 1250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-043-958 Supplier Gojo Industries Inc Supplier No. 882403
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Citrus ginger foam hand and shower wash is all-in-one body wash, shampoo, and hand soap that offers a refreshing blend of citrus extracts and ginger to enhance the shower experience.

Enriched with natural moisturizers and conditioners for skin and hair. USDA certified Biobased formulation. EcoLogo certified.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Body Wash
Color Light Yellow
Form Foam
For Use With (Equipment) PROVON™ ADX-12™ Dispenser
Size 1.25 L
Volume (Metric) 1250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.