Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ Provon Foaming Antimicrobial Handwash
SDP

Catalog No. 19044028 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
1250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-044-028 1250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-044-028 Supplier GOJO™ Supplier No. 518604
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Foaming antimicrobial handwash with moisturizers and light fruit fragrance.

  • Refill for PROVON FMX-12 Dispenser (GOJO 5160-06, sold separately).
  • For general-purpose handwashing with a foaming formulation.
  • Contains 0.3 percent PCMX.
  • Specially designed foaming formulations, enriched with vitamins and moisturizers, provide gentle cleansing experience along with a fresh, clean scent.
  • Push dispensers with viewing window for easy maintenance.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Soap
Color Orange
Form Foam
For Use With (Equipment) PROVON FMX-12 Push-Style Dispenser (Pk. of 4)
Size 1.25 L
Volume (Metric) 1250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.