Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High-Tech Conversions ProWorks™ High Dexterity Nitrile Gloves

Catalog No. 17444092
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-444-092 X-Large
17-444-091 Small
17-444-090 Medium
17-444-089 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-444-092 Supplier High-Tech Conversions Supplier No. GLN103FX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ProWorks™ High Dexterity Nitrile Gloves are powder free nitrile gloves that offer high dexterity and are suitable for industrial use.

  • Chemical resistant
  • Textured fingers
  • Meets FDA 21 CFR for food contact
  • 100/box, 10 boxes/case

Specifications

Certifications/Compliance FDA 21 CFR
Chlorinated No
Accelerator Free No
Applications Industrial, Laboratory
AQL 2.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Gloves
Size X-Large
Sterility Non-sterile
Thickness (English) 3 mil
Thickness (Metric) 0.0762 mm
Length (English) 9.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.