Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High-Tech Conversions ProWorks™ Polar Nite™ Nitrile Gloves

Catalog No. 17444095
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-444-095 Small
17-444-094 Medium
17-444-093 Large
17-444-096 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-444-095 Supplier High-Tech Conversions Supplier No. GLN133FS
Add to Cart
Add to Cart

ProWorks™ Polar Nite™ Nitrile Gloves are powder free, white nitrile exam gloves suitable for healthcare or laboratory use.

  • Excellent in areas when color coding is needed
  • Comfortable, same high quality of standard as blue gloves
  • Non-Sterile

Specifications

Certifications/Compliance ASTM D6319, FDA CFR Title 21 Food Additive Regulations, part 174-178
Chemotherapeutic Rated No
Chlorinated No
Accelerator Free No
Applications Healthcare, Laboratory
AQL 2.5
Color White
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 3 mil
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Small
Sterility Non-sterile
Thickness (English) 3 mil
Thickness (Metric) 0.07 mm
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model GL-N133FS
Quantity 1000/Cs.
Length (English) 9.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.