Learn More
Vector Laboratories VECTOR VIP Peroxidase (HRP) Substrate Kit 300 ml
SDP
Catalog No. NC9351198
Encompass
The Vector VIP Peroxidase (HRP) Substrate Kit produces intense violet (purple) reaction products and can be used as an alternative to DAB, or as a second color for multiple antigen labeling. Vector VIP chromogens also provides an excellent color contrast in pigmented tissues such as melanoma or retina. Vector VIP substrate can be used for both manual and automated staining methods. Stained sections should be dehydrated, cleared, and permanently mounted. The sensitivity of Vector VIP substrate is equivalent to Vector DAB Substrate.Vector VIP Substrate Kit reagents are provided in convenient dropper bottles, enough to prepare 300 ml of working solution. This product can also be used for blot-based protein detection.
Catalog No. NC9351198 Supplier Vector Laboratories Supplier No. SK4600
The Vector VIP Peroxidase (HRP) Substrate Kit produces intense violet (purple) reaction products and can be used as an alternative to DAB, or as a second color for multiple antigen labeling. Vector VIP chromogens also provides an excellent color contrast in pigmented tissues such as melanoma or retina. Vector VIP substrate can be used for both manual and automated staining methods. Stained sections should be dehydrated, cleared, and permanently mounted. The sensitivity of Vector VIP substrate is equivalent to Vector DAB Substrate.Vector VIP Substrate Kit reagents are provided in convenient dropper bottles, enough to prepare 300 ml of working solution. This product can also be used for blot-based protein detection.