Learn More
AdipoGen PROX1 human pAb
SDP
Supplier:  AdipoGen AG25T0104C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human PROX1. Source Rabbit. Applications IHC, WB. Lyophilized. Expression of PROX1 in blood endothelial cells induces expression of other lymphatic marker genes. Together with podoplanin, PROX1 can be used to reliably distinguish lympathic vessels from blood vessels. PROX1 is expressed in CNS, eye, pancreas, liver and heart and it is one of the most specific and reliable markers for lymphatic endothelial cells. The highly conserved C-terminal part of the homeobox transcription factor PROX1 was produced in E. coli. It was not tested for activity and can be used as positive control e.g. in Western analysis.
Catalog No. 50-256-3169