Learn More
AdipoGen PROX1 pAb
SDP
Supplier:  AdipoGen AG25T0105C050
Encompass_Preferred
Polyclonal Antibody. Recognizes human, chicken and horse PROX1. Source Rabbit. Applications IHC, WB. Lyophilized, 5 mM PBS, pH 7.2. Expression of PROX1 in blood endothelial cells induces expression of other lymphatic marker genes. Together with podoplanin, PROX1 can be used to reliably distinguish lympathic vessels from blood vessels. PROX1 is expressed in CNS, eye, pancreas, liver and heart, and it is one of the most specific and reliable markers for lymphatic endothelial cells.
Catalog No. 50-256-3168