Learn More
MSC Rochester Midland Corporation Proxi Encap, Encapsulation Carpet Cleaner, Odorless, 1 gal
Catalog No. 502837855
Encompass
The Proxi Encap Encapsulation Carpet Cleaner is a powerful, peroxide-based low-moisture maintenance cleaning formula designed to tackle heavy soil loads in carpets. This liquid concentrate effectively encapsulates dirt and odors for easy removal, leaving no sticky residue.
  • Encapsulates soil particles for quick and easy removal.
  • Low moisture cleaning, dries in 30 minutes.
  • Covers up to 5000 square feet per gallon.
  • Unique crystallizing polymer holds soil in suspension for dry vacuuming.
  • No sticky detergent residue.
  • Multi-method carpet cleaner.
  • Readily biodegradable and phosphate free.
  • Odorless formula.
  • Removes organic stains, odors, dirt, and soil.
  • Dilute 4-9 oz. per gallon of water.
  • Compatible with sprayer, pump, or encapsulation machine.
  • CRI approved.
Catalog No. 50-283-7855 Supplier MSC Supplier No. 36134369
The Proxi Encap Encapsulation Carpet Cleaner is a powerful, peroxide-based low-moisture maintenance cleaning formula designed to tackle heavy soil loads in carpets. This liquid concentrate effectively encapsulates dirt and odors for easy removal, leaving no sticky residue.
  • Encapsulates soil particles for quick and easy removal.
  • Low moisture cleaning, dries in 30 minutes.
  • Covers up to 5000 square feet per gallon.
  • Unique crystallizing polymer holds soil in suspension for dry vacuuming.
  • No sticky detergent residue.
  • Multi-method carpet cleaner.
  • Readily biodegradable and phosphate free.
  • Odorless formula.
  • Removes organic stains, odors, dirt, and soil.
  • Dilute 4-9 oz. per gallon of water.
  • Compatible with sprayer, pump, or encapsulation machine.
  • CRI approved.