Learn More
Honeywell™ Proximity Firefighting Gloves - Knit Wrist
Catalog No. 19815000
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-815-000 4X-Large
19-171-189D Medium
19-814-999 Large
19-051-530 X-Large
19-051-531 2X-Large
19-171-189H 3X-Large
19-051-529 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-815-000 Supplier Honeywell™ Supplier No. GLBPRRWA
Add to Cart
Add to Cart
Knit PBI and Aramid Fiber based aluminized material is the preferred shell for proximity turnout gear due to its flexibility, durability, and radiant heat reflectivity.
  • Knit PBI and Aramid Fiber based aluminized material is the preferred shell for Proximity turnout gear
  • Elkskin leather palm increases heat protection when gripping hot objects
  • Crosstech moisture barrier helps keep hands dry and comfortable
  • Aramid Fiber/Nomex™ lining for increased burn resistance
  • Supplemental wool felt insulation inside of the glove provides an additional layer of protection
  • Knit wrists helps prevent dirt and debris from entering the glove
Certifications
NFPA 1971-2007
TRUSTED_SUSTAINABILITY
Specifications
Back Material Aluminized PBI/Aramid Fiber
Certifications/Compliance NFPA 1971-2007
Cuff Material Leather Pull Tab
Coating Material Uncoated
Color Brown, Silver
Hand Specific
Product Type Proximity Firefighting Gloves
Size 4X-Large
Finger Style Full
Glove Cuff Color Gray
Glove Cuff Style Knit
Liner Material Nomex™/Aramid Fiber
Palm Type Elkhide Leather
Series GL-BPR-RWA
Thumb Style Wing