Learn More
Shelby™ Proximity Gloves
Catalog No. 19100344 Shop All Shelby Specialty Gloves Products
Click to view available options
Size:
Small
Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-100-344 Small
19-821-464 Large
19-100-343 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-100-344 Supplier Shelby™ Supplier No. 5200S
Add to Cart
Add to Cart
ARFF gloves offer reflective/radiant heat protection
  • Aluminized PBI™/Aramid Fiber knit ARFF glove offers reflective/radiant heat protection on the top, sides, and in between the fingers
  • Steamblock™ insulative pad on back of hand
  • Special gripping surfaces applied to the palm
Specifications
Certifications/Compliance NFPA 1976-2007
Material PBI, Aramid Fiber, Leather
Product Type Proximity Firefighting Gloves
Size Small
Glove Cuff Style Knit
Liner Material Thermal