Learn More
Pyramex™ Proximity™ Safety Eyewear
Catalog No. 19155877 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Anti-fog, Anti-scratch
Anti-fog, Anti-scratch, Anti-static
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor, Mirror
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint
19-155-877 Anti-fog Indoor/Outdoor, Mirror
19-155-872 Anti-fog, Anti-scratch, Anti-static Clear
19-155-873 Anti-fog, Anti-scratch Clear
19-155-874 Anti-fog, Anti-scratch, Anti-static Gray
19-155-875 Anti-fog, Anti-scratch Gray
19-155-876 Anti-fog, Anti-scratch, Anti-static Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-155-877 Supplier Pyramex™ Supplier No. SB9380ST
Add to Cart
Add to Cart
This item is not returnable. View return policy
9.5 base curve lens provides excellent side protection
  • Flame resistant foam padding
  • Ultra lightweight spectacle (26.3 grams)
  • Slim co-injected temples
  • Specifications
    Lens Coating Anti-fog
    Certifications/Compliance ANSI Z87.1 ,CSA Z94.3-07
    Certifications/Compliance (Europe) EN-166
    Lens Tint Indoor/Outdoor, Mirror
    Product Type Safety Spectacles
    Frame Color Black
    Weight (Metric) 26.3 g
    Material Polycarbonate
    Disposable Reusable
    Lens Material Polycarbonate
    UV Protection U6
    Frame Type Sealed