Learn More
Medchemexpress LLC HY-B1742 500mg Medchemexpress, Proxyphylline CAS:603-00-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1742500MG
Encompass_Preferred
Medchemexpress, HY-B1742 500mg Proxyphylline CAS:603-00-9 Proxyphylline is a methylxanthine derivative used as a cardiac stimulant, vasodilator and bronchodilator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2709