Learn More
Medchemexpress LLC HY-P0305 1mg Medchemexpress, PrP (106-126) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP03051MG
Encompass_Preferred
Medchemexpress, HY-P0305 1mg PrP (106-126) CAS: PrP (106-126) is a peptide corresponding to the prion protein (PrP) amyloidogenic region, and its biochemical properties resemble the infectious form of prion protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4109