Learn More
Aviva Systems Biology Corp PRPF4 Antibody - N-terminal region (ARP40542_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40542_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRPF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PRPF4. Alternate names: HPRP4, HPRP4P, PRP4, Prp4pApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57515