Learn More
ABclonal Technology PRPF4 Rabbit mAb
SDP
Catalog No. 5000007428
Encompass_Preferred
The protein encoded by this gene is part of a heteromeric complex that binds U4 U5 and U6 small nuclear RNAs and is involved in pre-mRNA splicing The encoded protein also is a mitotic checkpoint protein and a regulator of chemoresistance in human ovarian cancer Several transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-07428 Supplier ABclonal Technology Supplier No. A21132100UL
The protein encoded by this gene is part of a heteromeric complex that binds U4 U5 and U6 small nuclear RNAs and is involved in pre-mRNA splicing The encoded protein also is a mitotic checkpoint protein and a regulator of chemoresistance in human ovarian cancer Several transcript variants encoding different isoforms have been found for this gene