Learn More
Aviva Systems Biology Corp PRPF6 Antibody - C-terminal region (ARP40734_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40734_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PRPF6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PRPF6. Alternate names: ANT-1, TOM, ANT1, Prp6, hPrp6, U5-102K, C20orf14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53231