Learn More
Eurofins Discoverx ChemiScreen PRP/GPR10 Prolactin Releasing Peptide Receptor Stable Cell Line, 2 vials. (HTS057C)
Catalog No. 502005766
Encompass
The ChemiScreen PRP/GPR10 Prolactin Releasing Peptide Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prolactin releasing peptide) based activation of PRLHR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PRLHR alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of PRLHR, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5766 Supplier Eurofins Discoverx Supplier No. HTS057C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen PRP/GPR10 Prolactin Releasing Peptide Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prolactin releasing peptide) based activation of PRLHR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PRLHR alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of PRLHR, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.