Learn More
Eurofins Discoverx ChemiScreen PRP/GPR10 Prolactin-Releasing Peptide Receptor Membrane Preparation, 1 vial. (HTS057M)
Catalog No. 502005767
Encompass
The ChemiScreen PRP/GPR10 Prolactin-Releasing Peptide Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of PRLHR surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach or to screen for GPCR activity using GTPγS assays. Increased binding affinities or GPCR activation levels will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-5767 Supplier Eurofins Discoverx Supplier No. HTS057M
The ChemiScreen PRP/GPR10 Prolactin-Releasing Peptide Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of PRLHR surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach or to screen for GPCR activity using GTPγS assays. Increased binding affinities or GPCR activation levels will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.