Learn More
Eurofins Discoverx Ready-to-Assay PRP/GPR10 Prolactin-Releasing Peptide Receptor Frozen Cells, 2 Vials of Cells. (HTS057RTA)
Catalog No. 502005768
Encompass
The Ready-to-Assay PRP/GPR10 Prolactin-Releasing Peptide Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. prolactin releasing peptide) based activation of PRLHR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PRLHR alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of PRLHR, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5768 Supplier Eurofins Discoverx Supplier No. HTS057RTA
The Ready-to-Assay PRP/GPR10 Prolactin-Releasing Peptide Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. prolactin releasing peptide) based activation of PRLHR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PRLHR alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of PRLHR, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.