Learn More
Aviva Systems Biology Corp PRPS2 Antibody - middle region (ARP41448_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41448_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRPS2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human PRPS2. Alternate names: PRS II, PRSIIApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57861