Learn More
Aviva Systems Biology Corp Prpsap1 Antibody - N-terminal region (ARP56443_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56443_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Prpsap1. It was validated on Western Blot. . Alternate names: 5730409F23Rik, PAP39, RP23-193A16.1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-286-154