Learn More
Aviva Systems Biology Corp PRPSAP2 Antibody - middle region (ARP56446_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56446_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRPSAP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PRPSAP2. Alternate names: MGC117304, MGC126719, MGC126721, PAP41ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-155