Learn More
Aviva Systems Biology Corp PRR13 Antibody - middle region (ARP57260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRR13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRR13. Alternate names: DKFZp564J157, FLJ23818, TXR1ApplicationWB. Species reactivity: Cow, Dog, Human, Pig, Rabbit
Catalog No. 50-014-62113