Learn More
Aviva Systems Biology Corp PRR15 Antibody - middle region (ARP55745_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55745_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRR15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRR15. Alternate names: -ApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rat, Sheep
Catalog No. 50-286-075